Invitrogen
Manufacturer Code:PA5114785
Catalog # PIPA5114785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TNNT3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta TnTF; beta TnTF beta-TnTF Fast skeletal muscle troponin T fast skeletal muscle troponin T3 fast skeletal troponin T beta-1 isoform fTnT skeletal muscle fast-twitch TnT skeletal troponin T alpha-1 isoform Tnnt3 Tnt TNTF troponin T type 3 (skeletal fast) troponin T fast skeletal muscle troponin T3 fast skeletal type troponin T3 skeletal fast troponin-T3 skeletal fast; Beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fTnT; skeletal muscle fast-twitch TnT; TnTf; troponin T type 3 (skeletal, fast); Troponin T, fast skeletal muscle; troponin-T3, skeletal, fast
Gene Aliases: fTnT; TNNT3; TNTF
UniProt ID: (Human) P45378, (Mouse) O35585
Entrez Gene ID: (Human) 7140, (Mouse) 21957
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.