Proteintech
Manufacturer Code:119281AP150UL
Catalog # 50-172-7566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TMEFF2 Fusion Protein Ag2609 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen family 120, member 2; HPP1 TENB2 TMEFF2 Tomoregulin 2 TPEF TR TR 2; Hyperplastic polyposis protein 1; tomoregulin; Tomoregulin-2; TR-2; transmembrane protein TENB2; Transmembrane protein with EGF-like and two follistatin-like domains; transmembrane protein with EGF-like and two follistatin-like domains 2
Gene Aliases: 4832418D20Rik; 7630402F16Rik; CT120.2; HPP1; MNCb-1026; TENB2; TMEFF2; TPEF; TR; TR-2; UNQ178/PRO204
UniProt ID: (Human) Q9UIK5, (Mouse) Q9QYM9
Entrez Gene ID: (Human) 23671, (Mouse) 56363, (Rat) 363228
Molecular Function: extracellular matrix linker protein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.