R&D Systems
Manufacturer Code:IC7108G
Catalog # IC7108G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human TLR9 Asn64-Glu189 Accession # Q9NR96 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CD289; CD289 CD289 antigen toll-like receptor 9; Toll-like receptor 9
Gene Aliases: CD289; TLR9; UNQ5798/PRO19605
UniProt ID: (Human) Q9NR96
Entrez Gene ID: (Human) 54106
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.