Novus Biologicals
Manufacturer Code:NBP224655
Catalog # NBP224655
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acid 100-150 of human THEM2 was used as the immunogen for the antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA thioesterase 13; acyl-CoA thioesterase 13PNAS-27 EC 3.1.2.- HT012 hypothalamus protein HT012 MGC4961 THEM2 Thioesterase superfamily member 2acyl-coenzyme A thioesterase 13; Acyl-coenzyme A thioesterase 13; Acyl-coenzyme A thioesterase 13, N-terminally processed; Hotdog-fold thioesterase superfamily member 2; hypothalamus protein HT012; Palmitoyl-CoA hydrolase; THEM2; Thioesterase superfamily member 2
Gene Aliases: ACOT13; HT012; PNAS-27; THEM2
UniProt ID: (Human) Q9NPJ3
Entrez Gene ID: (Human) 55856
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.