Novus Biologicals
Manufacturer Code:NBP188146
Catalog # NBP188146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VNQPKRIENAKILIANTGMDTDKIKIFGSRVRVDSTAKVAEIEHAEKEKMKEKVERILKHGINCFINRQLIYNYPEQLFGAAGVMAIEHADFAGVERLALVTGGEIASTFDHPELVKLGSCKLIEEVMIGEDKLIHFSGV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT-beta; Cctb CCT-beta CCTBPRO163399D8.1 chaperonin containing t-complex polypeptide 1 beta subunit chaperonin containing t-complex polypeptide 1 subunit 2 chaperonin containing TCP1 subunit 2 (beta) MGC142074 MGC142076 T-complex protein 1 subunit beta T-complex protein 1 beta subunit TCP-1-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1, subunit 2 (beta); epididymis secretory sperm binding protein Li 100n; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; TCP-1-beta
Gene Aliases: 99D8.1; CCT-beta; CCT2; CCTB; HEL-S-100n; PRO1633; TCP-1-beta
UniProt ID: (Human) P78371
Entrez Gene ID: (Human) 10576
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.