R&D Systems
Manufacturer Code:MAB44061
Catalog # MAB44061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 401709 |
Immunogen | <EM>E. coli</EM>-derived recombinant human TCF-3/E2A Gly100-Glu500 Accession # NP_003191 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHb21; BHLHB21 bHLHb21TCF-3 Class B basic helix-loop-helix protein 21 E2A E2A immunoglobulin enhancer-binding factor E12/E47 E2AKappa-E2-binding factor Immunoglobulin enhancer-binding factor E12/E47 Immunoglobulin transcription factor 1 ITF1 ITF1Transcription factor ITF-1 Kappa-E2-binding factor MGC129647 MGC129648 Pan2 Tcfe2a Transcription factor 3 transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47) transcription factor E2-alpha VDIR VDR interacting repressor; Class B basic helix-loop-helix protein 21; helix-loop-helix protein HE47; Immunoglobulin enhancer-binding factor E12/E47; Immunoglobulin transcription factor 1; Kappa-E2-binding factor; negative vitamin D response element-binding protein; NOL1-TCF3 fusion; TCF-3; Transcription factor 3; transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47); transcription factor 3 variant 3; Transcription factor E2-alpha; Transcription factor ITF-1; VDR interacting repressor; vitamin D receptor-interacting repressor
Gene Aliases: AGM8; BHLHB21; E2A; E47; ITF1; TCF-3; TCF3; VDIR
UniProt ID: (Human) P15923
Entrez Gene ID: (Human) 6929
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.