Novus Biologicals
Manufacturer Code:NBP190421
Catalog # NBP190421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VDGESPNGSSDRGMKSLVNKMTVALKTKSVNVREKVISFIENTSTPVDRMSFNLPWPDRSCTERHVSSSDRVGKPYRGVKPVFSIGDEEEY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp667G062 HCV non-structural protein 4A-transactivated protein 1 HCV nonstructural protein 4A-transactivated protein 1 TBC1 domain family member 23 TBC1 domain family member 23; HCV non-structural protein 4A-transactivated protein 1; HCV nonstructural protein 4A-transactivated protein 1; TBC1 domain family member 23; TBC1 domain family, member 23
Gene Aliases: NS4ATP1; TBC1D23
UniProt ID: (Human) Q9NUY8
Entrez Gene ID: (Human) 55773
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.