Invitrogen
Manufacturer Code:PA577756
Catalog # PIPA577756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CHTKETLFMTGDMTHSE corresponding to amino acid residues 351-367 of rat Neurokinin Receptor 2 (NK2 ) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 7 transmembrane receptor; Neurokinin A receptor; NK-2 receptor; NK-2 receptor Tachykinin receptor 2 Neurokinin A receptor Neurokinin 2 receptor Substance-K receptor; NK-2R; SKR; substance K receptor; Substance-K receptor; Tachykinin 2 (substance K) receptor; tachykinin NK-2 receptor; Tachykinin receptor 2
Gene Aliases: Nk2r; Skr; Tac2r; Tacr2
UniProt ID: (Mouse) P30549, (Rat) P16610
Entrez Gene ID: (Mouse) 21337, (Rat) 25007
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.