Proteintech
Manufacturer Code:252701AP150UL
Catalog # 50-173-4826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neurokinin 2 receptor; Neurokinin A receptor; Neurokinin A receptor NK 2 receptor NK 2R NK2R NKNAR SKR Substance K receptor TAC2R tachykinin receptor 2 TACR2; NK-2 receptor; NK-2R; seven transmembrane helix receptor; SKR; substance K receptor; Substance-K receptor; Tachykinin receptor 2
Gene Aliases: NK2R; NKNAR; SKR; TAC2R; TACR2
UniProt ID: (Human) P21452
Entrez Gene ID: (Human) 6865
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.