Invitrogen
Manufacturer Code:PA5113726
Catalog # PIPA5113726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SYNJ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A930006D20Rik AA675315 inositol 5&prime-phosphatase (synaptojanin 1) inositol polyphosphate-5-phosphatase G INPP5G KIAA0910 mKIAA0910 PARK20 phosphoinositide 5-phosphatase Synaptic inositol 145-trisphosphate 5-phosphatase 1 synaptic inositol-1 synaptic inositol-145-trisphosphate 5-phosphatase 1 synaptojanin 1 Synaptojanin-1 synaptojanin-1 polyphosphoinositide phosphatase SYNJ1; inositol 5'-phosphatase (synaptojanin 1); inositol polyphosphate-5-phosphatase G; phosphoinositide 5-phosphatase; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 1; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 1; Synaptojanin-1; synaptojanin-1, polyphosphoinositide phosphatase
Gene Aliases: INPP5G; KIAA0910; PARK20; SYNJ1
UniProt ID: (Human) O43426
Entrez Gene ID: (Human) 8867
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.