Invitrogen
Manufacturer Code:PA518990
Catalog # PIPA518990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DETNKGFQPDEK) corresponding to the C-terminus amino acids of Slc10a2* |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Apical sodium-dependent bile acid transporter; ASBT; IBAT; Ileal Na(+)/bile acid cotransporter; Ileal sodium-dependent bile acid transporter; Ileal sodium/bile acid cotransporter; Na(+)-dependent ileal bile acid transporter; Slc10a2 solute carrier family 10 (sodium/bile acid cotransporter family) member 2 ASBT ISBT ileal apical sodium-dependent bile acid transporter ileal sodium-dependent bile acid transporter solute carrier family 10 member 2; Sodium/taurocholate cotransporting polypeptide, ileal; Solute carrier family 10 member 2
Gene Aliases: ASBT; ISBT; Ntcp2; Slc10a2
UniProt ID: (Mouse) P70172
Entrez Gene ID: (Mouse) 20494
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.