Invitrogen
Manufacturer Code:PA568340
Catalog # PIPA568340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human SV2C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1054; Synaptic vesicle glycoprotein 2C; Synaptic vesicle protein 2C
Gene Aliases: 4930527L09Rik; AI841971; KIAA1054; SV2C
UniProt ID: (Human) Q496J9, (Mouse) Q69ZS6, (Rat) Q9Z2I6
Entrez Gene ID: (Human) 22987, (Mouse) 75209, (Rat) 29643
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.