Novus Biologicals
Manufacturer Code:NB10041402
Catalog # NB10041402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-TVELYPSDKRTTA corresponding to internal region according to NP_055664.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0736SV2 synaptic vesicle glycoprotein 2A; Synaptic vesicle glycoprotein 2A
Gene Aliases: KIAA0736; PSEC0174; SV2; SV2A
UniProt ID: (Human) Q7L0J3
Entrez Gene ID: (Human) 9900
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.