Abnova
Manufacturer Code:H00010809K
Catalog # 89-207-350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human STARD10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human STARD10 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Antigen NY-CO-28; CGI-52 MGC14401 NY-CO-28 PCTP2 SDCCAG28; PCTP-L; PCTP-like protein; Serologically defined colon cancer antigen 28; StAR-related lipid transfer (START) domain containing 10; StAR-related lipid transfer domain containing 10; StAR-related lipid transfer protein 10; StARD10; START domain containing 10; START domain-containing protein 10
Gene Aliases: CGI-52; NY-CO-28; PCTP2; SDCCAG28; STARD10
UniProt ID: (Human) Q9Y365
Entrez Gene ID: (Human) 10809
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.