Invitrogen
Manufacturer Code:PA543390
Catalog # PIPA543390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SRPRB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein APMCF1; Signal recognition particle receptor B subunit APMCF1; Signal recognition particle receptor subunit beta; signal recognition particle receptor subunit beta pseudogene; signal recognition particle receptor, B subunit; signal recognition particle receptor, beta subunit; SR-beta; SRP receptor beta subunit
Gene Aliases: AA409543; Ab2-417; APMCF1; Ba1-667; Cc1-8; PSEC0230; SRPRB; wu:fk57h10; zgc:111820; zgc:92746
UniProt ID: (Human) Q9Y5M8, (Rat) Q4FZX7, (Mouse) P47758
Entrez Gene ID: (Human) 58477, (Rabbit) 100337982, (Dog) 102154222, (Horse) 100629957, (Rat) 300965, (Mouse) 20818, (Zebrafish) 436845
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.