Novus Biologicals
Manufacturer Code:NBP213380
Catalog # NBP213380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: SALRSLSNATIINEEVLNAMLKEVCTALLEADVNIKLVKQLRENVKSAID LEEMASGLNKRKMIQHAVFKELVKLVDPGVKAWTPTKGKQNV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Signal recognition particle 54 kDa protein; signal recognition particle 54 kDa protein signal recognition particle 54kD signal recognition particle 54kDa; signal recognition particle 54kD; signal recognition particle 54kDa; SRP54
Gene Aliases: SRP54
UniProt ID: (Human) P61011
Entrez Gene ID: (Human) 6729
Molecular Function: G-protein RNA binding protein enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.