Novus Biologicals
Manufacturer Code:NBP294184100UL
Catalog # NB022925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-136 of human SRP14 (NP_003125.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa Alu RNA-binding protein; ALURBP MGC1432618 kDa Alu RNA-binding protein signal recognition particle 14 kDa protein signal recognition particle 14kD (homologous Alu RNA-binding protein) signal recognition particle 14kDa (homologous Alu RNA binding protein); homologous Alu RNA binding protein; Signal recognition particle 14 kDa protein; signal recognition particle 14kDa (homologous Alu RNA binding protein); SRP14
Gene Aliases: ALURBP; SRP14
UniProt ID: (Human) P37108
Entrez Gene ID: (Human) 6727
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.