Proteintech
Manufacturer Code:115281AP150UL
Catalog # 50-172-7238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SRP14 Fusion Protein Ag2099 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa Alu RNA binding protein ALURBP SRP14; 18 kDa Alu RNA-binding protein; homologous Alu RNA binding protein; Signal recognition particle 14 kDa protein; signal recognition particle 14kDa (homologous Alu RNA binding protein); SRP14
Gene Aliases: ALURBP; SRP14
UniProt ID: (Human) P37108
Entrez Gene ID: (Human) 6727, (Rat) 296076
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.