Novus Biologicals
Manufacturer Code:NBP154446SS
Catalog # NBP154446SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Gel Shift (GS) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D2 |
Immunogen | Human SREBP2 protein (amino acids 48-403). [UniProt# Q12772] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHd2; BHLHD2 bHLHd2sterol regulatory element-binding protein 2 Class D basic helix-loop-helix protein 2 SREBP2 SREBP2SREBP-2 sterol regulatory element binding transcription factor 2 Sterol regulatory element-binding transcription factor 2; Class D basic helix-loop-helix protein 2; Processed sterol regulatory element-binding protein 2; SREBP-2; Sterol regulatory element-binding protein 2; Sterol regulatory element-binding transcription factor 2; Transcription factor SREBF2
Gene Aliases: BHLHD2; SREBF2; SREBP-2; SREBP2
UniProt ID: (Human) Q12772
Entrez Gene ID: (Human) 6721
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.