Abnova
Manufacturer Code:PAB24187
Catalog # 89-125-894
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SPNS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Protein Aliases: HSpin1; Protein spinster homolog 1; spinster homolog 1; Spinster-like protein 1; SPNS sphingolipid transporter 1 (putative)
Gene Aliases: HSpin1; LAT; nrs; PP2030; PP20300; SPIN1; SPINL; SPNS1
UniProt ID: (Human) Q9H2V7
Entrez Gene ID: (Human) 83985
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.