Novus Biologicals
Manufacturer Code:NBP274312B
Catalog # NBP274312B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E4 |
Immunogen | Recombinant protein fragment corresponding to amino acids 2-64 of human SPCS2 (NP_055567) produced in E.coli. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: MGC117366 signal peptidase complex subunit 2 signal peptidase complex subunit 2 homolog (S. cerevisiae) SPC25; Microsomal signal peptidase 25 kDa subunit; signal peptidase 25kDa subunit; Signal peptidase complex subunit 2; signal peptidase complex subunit 2 homolog; SPase 25 kDa subunit
Gene Aliases: KIAA0102; SPC25; SPCS2
UniProt ID: (Human) Q15005
Entrez Gene ID: (Human) 9789
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.