Proteintech
Manufacturer Code:110641AP150UL
Catalog # 50-172-6844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SOX2 Fusion Protein Ag1530 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANOP3 MCOPS3 SOX2 Transcription factor SOX 2; sex determining region Y-box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY-box containing gene 2; SRY-related HMG-box gene 2; Transcription factor SOX-2; transcription factor SOX2
Gene Aliases: ANOP3; cb236; lcc; MCOPS3; RGD1565646; Sox-2; SOX2; soxp; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389
UniProt ID: (Human) P48431, (Mouse) P48432, (Zebrafish) Q6P0E1
Entrez Gene ID: (Human) 6657, (Rat) 499593, (Mouse) 20674, (Zebrafish) 378723
Molecular Function: HMG box transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.