Novus Biologicals
Manufacturer Code:NBP303762100UL
Catalog # NB095611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 546-635 of human SMVT/SLC5A6 (NP_066918.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)-dependent multivitamin transporter; Na(+)-dependent multivitamin transporter SMVTNa+-dependent multivitamin transporter sodium-dependent multivitamin transporter solute carrier family 5 (sodium-dependent vitamin transporter) member 6 Solute carrier family 5 member 6; Na+-dependent multivitamin transporter; Sodium-dependent multivitamin transporter; solute carrier family 5 (sodium-dependent vitamin transporter), member 6; solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6; Solute carrier family 5 member 6
Gene Aliases: SLC5A6; SMVT
UniProt ID: (Human) Q9Y289
Entrez Gene ID: (Human) 8884
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.