Novus Biologicals
Manufacturer Code:NBP220400
Catalog # NBP220400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human SLITRK5. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA364G4.2 FLJ58374 KIAA0918 leucine-rich repeat-containing protein 11 LRRC11 SLIT and NTRK-like family member 5 SLIT and NTRK-like protein 5; leucine rich repeat containing 11; Leucine-rich repeat-containing protein 11; SLIT and NTRK-like family, member 5; SLIT and NTRK-like protein 5; slit and trk like gene 5
Gene Aliases: bA364G4.2; KIAA0918; LRRC11; SLITRK5
UniProt ID: (Human) O94991
Entrez Gene ID: (Human) 26050
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.