R&D Systems
Manufacturer Code:AF2587SP
Catalog # AF2587SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human SLITRK5 Glu39-Val665 Accession # O94991 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bA364G4.2 FLJ58374 KIAA0918 leucine-rich repeat-containing protein 11 LRRC11 SLIT and NTRK-like family member 5 SLIT and NTRK-like protein 5; leucine rich repeat containing 11; Leucine-rich repeat-containing protein 11; SLIT and NTRK-like family, member 5; SLIT and NTRK-like protein 5; slit and trk like gene 5
Gene Aliases: bA364G4.2; KIAA0918; LRRC11; SLITRK5
UniProt ID: (Human) O94991
Entrez Gene ID: (Human) 26050
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.