Invitrogen
Manufacturer Code:PA542453
Catalog # PIPA542453
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLCO2B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: multispecific organic anion transporter 1; OATP-B; OATP-RP2; Oatp2b1; Organic anion transporter B; Organic anion transporter moatp1; Organic anion transporter polypeptide-related protein 2; organic anion transporting polypeptide 2b1; organic anion-transporting polypeptide 2B1; SLCO/OATP2B1; solute carrier anion transporting family member 2b1; solute carrier family 21 (organic anion transporter), member 9; Solute carrier family 21 member 9; Solute carrier organic anion transporter family member 2B1; solute carrier organic anion transporter family, member 2B1
Gene Aliases: AI060904; AI852653; im:7158298; KIAA0880; moat1; OATP-B; OATP2B1; OATPB; SLC21A9; SLCO2B1; wu:fi40d02; zgc:123236
UniProt ID: (Human) O94956, (Mouse) Q8BXB6, (Rat) Q9JHI3
Entrez Gene ID: (Rabbit) 100353580, (Human) 11309, (Dog) 485192, (Horse) 100009692, (Bovine) 317704, (Mouse) 101488, (Guinea pig) 100732070, (Rat) 140860, (Zebrafish) 792084
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.