Novus Biologicals
Manufacturer Code:NBP29314020UL
Catalog # NB028169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human SLCO1C1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OAT-RP-5; OATP-14; OATP-F; OATP1 OATP14Thyroxine transporter OATP1C1Solute carrier family 21 member 14 OATPFOAT-RP-5 OATP-FOrganic anion-transporting polypeptide 14 OATPRP5 Organic anion transporter F Organic anion transporter polypeptide-related protein 5 organic anion transporting polypeptide 14 SLC21A14OATP-14 solute carrier family 21 (organic anion transporter) member 14 solute carrier organic anion transporter family member 1C1 solute carrier organic anion transporter family member 1C1; Organic anion transporter F; Organic anion transporter polypeptide-related protein 5; organic anion transporting polypeptide 14; Organic anion-transporting polypeptide 14; solute carrier family 21 (organic anion transporter), member 14; Solute carrier family 21 member 14; Solute carrier organic anion transporter family member 1C1; solute carrier organic anion transporter family, member 1C1; Thyroxine transporter
Gene Aliases: OATP-F; OATP1; OATP14; OATP1C1; OATPF; OATPRP5; SLC21A14; SLCO1C1
UniProt ID: (Human) Q9NYB5
Entrez Gene ID: (Human) 53919
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.