Invitrogen
Manufacturer Code:PA542455
Catalog # PIPA542455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC7A8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4F2-LC5; cationic amino acid transporter y+ system; glycoprotein-associated amino acid transporter LAT2; hLAT2; integral membrane protein E16H; L-type amino acid transporter 2; Large neutral amino acids transporter small subunit 2; mLAT2; SLC7A8; Solute carrier family 7 (amino acid transporter light chain L system) member 8 LAT2 LPI-PC1; solute carrier family 7 (amino acid transporter light chain, L system), member 8; solute carrier family 7 (amino acid transporter, L-type), member 8; solute carrier family 7 (cationic amino acid transporter, y+ system), member 8; Solute carrier family 7 member 8; solute carrier family 8 (cationic amino acid transporter, y+ system), member 7
Gene Aliases: 4F2LC-5; AA408822; LAT2; Lat4; LPI-PC1; SLC7A8
UniProt ID: (Human) Q9UHI5, (Rabbit) Q9N1Q4, (Mouse) Q9QXW9, (Rat) Q9WVR6
Entrez Gene ID: (Human) 23428, (Dog) 490608, (Rabbit) 100009408, (Bovine) 536818, (Horse) 100052453, (Mouse) 50934, (Guinea pig) 100717482, (Rat) 84551
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.