Abnova
Manufacturer Code:PAB14941
Catalog # 89-117-361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 1-50 of mouse Slc7a11. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc7a11.
For Research Use Only
Protein Aliases: 9930009M05Rik; AI451155; Amino acid transport system xc-; cysteine/glutamate transporter; Cystine/glutamate transporter; sodium independent anionic amino acid transport system; Solute carrier family 7 member 11; sut; xCT
Gene Aliases: 9930009M05Rik; AI451155; Slc7a11; sut; xCT
UniProt ID: (Mouse) Q9WTR6
Entrez Gene ID: (Mouse) 26570
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.