Invitrogen
Manufacturer Code:PA539460
Catalog # PIPA539460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human SLC6A8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: choline transporter; CHOT1; creatine transporter 1; creatine transporter SLC6A8 variant D; creatine transporter, solute carrier family 6, member 8; CT1; Sodium- and chloride-dependent creatine transporter 1; solute carrier family 6 (neurotransmitter transporter), member 8; solute carrier family 6 (neurotransmitter transporter, creatine), member 8; Solute carrier family 6 member 8; solute carrier family 6 neurotransmitter transporter creatine member 8 CRT CRTR CT1 MGC87396 creatine transporter 1 creatine transporter SLC6A8 sodium- and chloride-dependent creatine transporter 1 solute carrier family 6 member 8; solute carrier family 6, member 8
Gene Aliases: AA589632; CCDS1; CHOT1; CHT1; Creat; CRT; CRTR; CT1; CTR5; SLC6A8
UniProt ID: (Human) P48029, (Rat) P28570, (Mouse) Q8VBW1
Entrez Gene ID: (Human) 6535, (Rat) 50690, (Mouse) 102857
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.