Novus Biologicals
Manufacturer Code:NBP186277
Catalog # NBP186277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GFRATQRYDDCFSTNILTLINGFDLPEGNVTQENFVDMQQRCNASDPAAYAQLVFQTCDINAFLSEAVEGT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B0AT1 FLJ20680 FLJ34635 HND sodium-dependent amino acid transporter system B0 sodium-dependent neutral amino acid transporter B(0)AT1 solute carrier family 6 (neurotransmitter transporter) member 19 solute carrier family 6 (neutral amino acid transporter) member 19 Solute carrier family 6 member 19 System B(0) neutral amino acid transporter AT1 system B0 neutral amino acid transporter; sodium-dependent amino acid transporter system B0; Sodium-dependent neutral amino acid transporter B(0)AT1; solute carrier family 6 (neurotransmitter transporter), member 19; solute carrier family 6 (neutral amino acid transporter), member 19; Solute carrier family 6 member 19; System B(0) neutral amino acid transporter AT1; system B0 neutral amino acid transporter
Gene Aliases: B0AT1; HND; SLC6A19
UniProt ID: (Human) Q695T7
Entrez Gene ID: (Human) 340024
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.