Invitrogen
Manufacturer Code:PA577451
Catalog # PIPA577451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KVEEDGTLEREQWTNK corresponding to amino acids residues 23-38 of rat GABA Transporter 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma-aminobutyric acid (GABA-A) transporter 3; GAT-2; GAT-3; SLC6A13; Sodium- and chloride-dependent GABA transporter 2; Sodium- and chloride-dependent GABA transporter 3; solute carrier family 6 (neurotransmitter transporter, GABA), member 13; Solute carrier family 6 member 13
Gene Aliases: Gabt2; Gabt3; GAT-2; Gat-3; Gat2; GAT3; Slc6a13
UniProt ID: (Mouse) P31649, (Rat) P31646
Entrez Gene ID: (Mouse) 14412, (Rat) 171163
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.