Invitrogen
Manufacturer Code:PA577453
Catalog # PIPA577453
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GEMMDQKDKDQVKD corresponding to amino acid residues 22-35 of rat Betaine/GABA Transporter 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Betaine/GABA transporter 1; betaine/GABA Transporter 1 (BGT-1); GABA transporter 1; gamma-aminobutyric acid (GABA-A) transporter 2; GAT-2; Na(+)/Cl(-) betaine/GABA transporter; Sodium- and chloride-dependent betaine transporter; sodium- and chloride-dependent GABA transporter 2; solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12; Solute carrier family 6 member 12; vesicular GABA transporter
Gene Aliases: BGT1; Gabt2; Gat1; GAT2; RNU28927; Slc6a12; VGAT
UniProt ID: (Rat) P48056
Entrez Gene ID: (Mouse) 14411, (Rat) 50676
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.