Invitrogen
Manufacturer Code:PA585766
Catalog # PIPA585766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of mouse GABA Transporter 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A730043E01 antibody GABATHG antibody GABATR antibody GAT-1 antibody Gabt antibody Gabt1 antibody Gat1 antibody Slc6a1 antibody XT1 antibody Xtrp1 antibody solute carrier family 6 (neurotransmitter transporter GABA) member 1 antibody; GABA transporter protein; gamma-aminobutyric acid (GABA-A) transporter 1; GAT-1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; Solute carrier family 6 member 1
Gene Aliases: A730043E01; GABATHG; GABATR; Gabt; Gabt1; GAT-1; Gat1; Slc6a1; XT-1; Xtrp1
UniProt ID: (Rat) P23978, (Mouse) P31648
Entrez Gene ID: (Rat) 79212, (Mouse) 232333
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.