R&D Systems
Manufacturer Code:FAB8398N100UG
Catalog # FAB8398N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 903502 |
Immunogen | NS0 mouse myeloma cell line transfected with human SLC5A8/SMCT1 Met1-Leu610 Accession # NP_666018 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: AIT AITSolute carrier family 5 member 8 Apical iodide transporter Electrogenic sodium monocarboxylate cotransporter MGC125354 SMCT SMCTSMCT1 Sodium iodide-related cotransporter sodium-coupled monocarboxylate transporter 1 solute carrier family 5 (iodide transporter) member 8; Apical iodide transporter; Electrogenic sodium monocarboxylate cotransporter; SLC5A8; Sodium iodide-related cotransporter; Sodium-coupled monocarboxylate transporter 1; solute carrier family 5 (iodide transporter), member 8; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 8; Solute carrier family 5 member 8
Gene Aliases: AIT; SLC5A8; SMCT; SMCT1
UniProt ID: (Human) Q8N695
Entrez Gene ID: (Human) 160728
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.