Invitrogen
Manufacturer Code:PA542514
Catalog # PIPA542514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC5A8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apical iodide transporter; apical iodide transporter , putative; Electrogenic sodium monocarboxylate cotransporter; SLC5A8; Sodium iodide-related cotransporter; Sodium-coupled monocarboxylate transporter 1; solute carrier family 5 (iodide transporter), member 8; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 8; Solute carrier family 5 member 8
Gene Aliases: AIT; RGD1564146; SLC5A8; SMCT; SMCT1
UniProt ID: (Human) Q8N695, (Mouse) Q8BYF6
Entrez Gene ID: (Rabbit) 100339808, (Human) 160728, (Mouse) 216225, (Rat) 500820
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.