Proteintech
Manufacturer Code:104531AP150UL
Catalog # 50-172-6320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC45A2 Fusion Protein Ag0699 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1A1 AIM1 MATP Melanoma antigen AIM1 Protein AIM 1 SHEP5 SLC45A2; B/AIM-1-like protein; dominant brown; Melanoma antigen AIM1; membrane associated transporter protein; Membrane-associated transporter protein; Protein AIM-1; Protein underwhite; Solute carrier family 45 member 2; solute carrier family 45, member 2; underwhite
Gene Aliases: 1A1; Aim-1; AIM1; blanc-sale; bls; Dbr; MATP; OCA4; SHEP5; SLC45A2; uw
UniProt ID: (Human) Q9UMX9, (Mouse) P58355
Entrez Gene ID: (Human) 51151, (Rat) 310152, (Mouse) 22293
Molecular Function: carbohydrate transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.