Abnova
Manufacturer Code:PAB28364
Catalog # 89-129-867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acids at internal region of human SLC44A2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyconal antibody raised against synthetic peptide of SLC44A2.
For Research Use Only
Protein Aliases: Choline transporter-like protein 2; CTL2; DKFZp666A071; FLJ44586; PP1292; solute carrier family 44 (choline transporter), member 2; Solute carrier family 44 member 2; solute carrier family 44, member 2; testicular tissue protein Li 47
Gene Aliases: CTL2; PP1292; PSEC0210; SLC44A2
UniProt ID: (Human) Q8IWA5
Entrez Gene ID: (Human) 57153
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.