Invitrogen
Manufacturer Code:PA557187
Catalog # PIPA557187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC38A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amino acid transporter 2; Amino acid transporter A2; Protein 40-9-1; Sodium-coupled neutral amino acid transporter 2; Solute carrier family 38 member 2; Solute carrier family 38 member 2 ATA2 SAT2 SNAT2 PRO1068; solute carrier family 38, member 2; System A amino acid transporter 2; System A transporter 1; System N amino acid transporter 2
Gene Aliases: ATA2; KIAA1382; PRO1068; SAT2; SLC38A2; SNAT2
UniProt ID: (Human) Q96QD8
Entrez Gene ID: (Human) 54407
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.