Novus Biologicals
Manufacturer Code:NBP2835540.1ML
Catalog # NB031483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human SLC38A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amino acid transporter 2; Amino acid transporter A2; Amino acid transporter A2 ata2 ATA2System A transporter 1 KIAA1382System N amino acid transporter 2 pro1068 Protein 40-9-1 sat2 SAT2PRO1068 snat2 SNAT2amino acid transporter 2 sodium-coupled neutral amino acid transporter 2 Solute carrier family 38 member 2 solute carrier family 38 member 2 System A amino acid transporter 2; Protein 40-9-1; Sodium-coupled neutral amino acid transporter 2; Solute carrier family 38 member 2; solute carrier family 38, member 2; System A amino acid transporter 2; System A transporter 1; System N amino acid transporter 2
Gene Aliases: ATA2; KIAA1382; PRO1068; SAT2; SLC38A2; SNAT2
UniProt ID: (Human) Q96QD8
Entrez Gene ID: (Human) 54407
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.