Abnova
Manufacturer Code:H00054407K
Catalog # 89-201-723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC38A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC38A2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: amino acid transporter 2; Amino acid transporter A2; ATA2 KIAA1382 PRO1068 SAT2 SNAT2; Protein 40-9-1; Sodium-coupled neutral amino acid transporter 2; Solute carrier family 38 member 2; solute carrier family 38, member 2; System A amino acid transporter 2; System A transporter 1; System N amino acid transporter 2
Gene Aliases: ATA2; KIAA1382; PRO1068; SAT2; SLC38A2; SNAT2
UniProt ID: (Human) Q96QD8
Entrez Gene ID: (Human) 54407
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.