Invitrogen
Manufacturer Code:PA569289
Catalog # PIPA569289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse SLC35F3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Putative thiamine transporter SLC35F3; Solute carrier family 35 member F3; solute carrier family 35, member F3
Gene Aliases: B230375D17Rik; SLC35F3
UniProt ID: (Human) Q8IY50, (Mouse) Q1LZI2
Entrez Gene ID: (Guinea pig) 100714042, (Human) 148641, (Rabbit) 100341547, (Horse) 100059412, (Bovine) 538365, (Dog) 488963, (Rat) 679321, (Mouse) 210027
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.