Invitrogen
Manufacturer Code:MA531330
Catalog # PIMA531330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL0215 |
Immunogen | Recombit protein corresponding to Human SLC27A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACSB ACSVL6 FACVL3 FATP5 FLJ22987 VLACSR VLCS-H2 VLCSH2; BA-CoA ligase; BACS; Bile acid-CoA ligase; Bile acyl-CoA synthetase; Cholate--CoA ligase; FATP-5; Fatty acid transport protein 5; Fatty-acid-coenzyme A ligase, very long-chain 3; solute carrier family 27 (fatty acid transporter), member 5; Solute carrier family 27 member 5; Very long-chain acyl-CoA synthetase homolog 2; Very long-chain acyl-CoA synthetase-related protein; VLACS-related; VLCS-H2
Gene Aliases: ACSB; ACSVL6; BACS; BAL; FACVL3; FATP-5; FATP5; SLC27A5; VLACSR; VLCS-H2; VLCSH2
UniProt ID: (Human) Q9Y2P5
Entrez Gene ID: (Human) 10998
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.