Invitrogen
Manufacturer Code:PA570402
Catalog # PIPA570402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human SLC25A6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAC3 ANT3 ANT3Y ADPATP carrier protein ADPATP carrier protein 3 ADPATP carrier protein liver ADP/ATP translocase 3 ADP/ATP translocator of liver ANT 2 ANT 3 adenine nucleotide translocator 3 solute carrier family 25 member 6; adenine nucleotide translocator 2; Adenine nucleotide translocator 3; adenine nucleotide translocator 3 (liver); ADP,ATP carrier protein 3; ADP,ATP carrier protein, isoform T2; ADP,ATP carrier protein, liver; ADP/ATP translocase 3; ADP/ATP translocase 3, N-terminally processed; ADP/ATP translocator of liver; ANT 2; ANT 3; mitochondrial ADP/ATP carrier protein; mitochondrial solute carrier family 25 member 6; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6; Solute carrier family 25 member 6; solute carrier family 25, member A6
Gene Aliases: AAC3; ANT; ANT 2; ANT 3; ANT3; ANT3Y; CDABP0051; si:dkey-21o13.4; SLC25A6; wu:fj78b08
UniProt ID: (Human) P12236, (Pig) Q6QRN9, (Bovine) P32007
Entrez Gene ID: (Human) 293, (Pig) 403155, (Dog) 480830, (Bovine) 282480, (Sheep) 100147789, (Zebrafish) 566370
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.