Novus Biologicals
Manufacturer Code:NBP294907100UL
Catalog # NB024320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human SLC23A2 (NP_976072.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSVCT2; KIAA0238SLC23A1 Na(+)/L-ascorbic acid transporter 2 Nucleobase transporter-like 1 protein Sodium-dependent vitamin C transporter 2 sodium-dependent vitamin C transporter-2 solute carrier family 23 (nucleobase transporters) member 1 solute carrier family 23 (nucleobase transporters) member 2 solute carrier family 23 member 2 SVCT2NBTL1 Yolk sac permease-like molecule 2 YSPL2hSVCT2; Na(+)/L-ascorbic acid transporter 2; Nucleobase transporter-like 1 protein; Sodium-dependent vitamin C transporter 2; sodium-dependent vitamin C transporter-2; solute carrier family 23 (ascorbic acid transporter), member 2; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; Solute carrier family 23 member 2; testicular secretory protein Li 48; Yolk sac permease-like molecule 2
Gene Aliases: hSVCT2; KIAA0238; NBTL1; SLC23A1; SLC23A2; SVCT2; YSPL2
UniProt ID: (Human) Q9UGH3
Entrez Gene ID: (Human) 9962
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.