Invitrogen
Manufacturer Code:MA537983
Catalog # PIMA537983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2285 |
Immunogen | A synthesized peptide derived from human SLC22A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-Oct EMT EMT organic cation transporter 3 EMTH Extraneuronal monoamine transporter OCT3 Orct3 Organic cation transporter 3 Slc22a3 Slca22a3 solute carrier family 22 (extraneuronal monoamine transporter) member 3 solute carrier family 22 (organic cation transporter) member 3 Solute carrier family 22 member 3 solute carrier family 22 member 3; EMT; EMT organic cation transporter 3; Extraneuronal monoamine transporter; Organic cation transporter 3; solute carrier family 22 (extraneuronal monoamine transporter), member 3; solute carrier family 22 (organic cation transporter), member 3; Solute carrier family 22 member 3; solute carrier family 22, member 3
Gene Aliases: EMT; EMTH; OCT3; Orct3; SLC22A3; Slca22a3
UniProt ID: (Human) O75751, (Mouse) Q9WTW5
Entrez Gene ID: (Human) 6581, (Mouse) 20519
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.