Proteintech
Manufacturer Code:249011AP150UL
Catalog # 50-173-4606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC22A23 Fusion Protein Ag19036 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOCT2 C6orf85 SLC22A23; Ion transporter protein; Solute carrier family 22 member 23; solute carrier family 22, member 23
Gene Aliases: 3110004L20Rik; C6orf85; Nritp; SLC22A23
UniProt ID: (Human) A1A5C7, (Mouse) Q3UHH2, (Rat) Q9QZG1
Entrez Gene ID: (Human) 63027, (Mouse) 73102, (Rat) 64559
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.