R&D Systems
Manufacturer Code:MAB6547SP
Catalog # MAB6547SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 640438 |
Immunogen | NS0 mouse myeloma cell line transfected with human SLC-22A2 Accession # O15244 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hOCT2; hOCT2 MGC32628 OCT2 OCT2Organic cation transporter 2 solute carrier family 22 (organic cation transporter) member 2 solute carrier family 22 member 2; Organic cation transporter 2; solute carrier family 22 (organic cation transporter), member 2; Solute carrier family 22 member 2
Gene Aliases: OCT2; SLC22A2
UniProt ID: (Human) O15244
Entrez Gene ID: (Human) 6582
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.