Abnova
Manufacturer Code:H00006582M02
Catalog # 89-105-463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E3 |
Immunogen | SLC22A2 (AAH39899261 a.a. &bsim 370 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SLC22A2.
For Research Use Only
Protein Aliases: hOCT2; MGC32628; OCT2; Organic cation transporter 2; solute carrier family 22 (organic cation transporter), member 2; Solute carrier family 22 member 2
Gene Aliases: OCT2; SLC22A2
UniProt ID: (Human) O15244
Entrez Gene ID: (Human) 6582
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.