Invitrogen
Manufacturer Code:PA542477
Catalog # PIPA542477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC22A11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Organic anion transporter 4; solute carrier family 22 (organic anion/cation transporter), member 11; Solute carrier family 22 (organic anion/urate transporter) member 11 MGC34282 OAT4 hOAT4; solute carrier family 22 (organic anion/urate transporter), member 11; Solute carrier family 22 member 11
Gene Aliases: hOAT4; OAT4; SLC22A11
UniProt ID: (Human) Q9NSA0
Entrez Gene ID: (Human) 55867
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.